Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATGTGCACTACTGAAGAGCTTCT[A/G]GTATTAGACTAGAAAAAGAAAAGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606409 | ||||||||||||||||||||
Literature Links: |
ITCH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ITCH - itchy E3 ubiquitin protein ligase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257137.2 | 189 | Intron | NP_001244066.1 | |||
NM_001257138.2 | 189 | Intron | NP_001244067.1 | |||
NM_001324197.1 | 189 | Intron | NP_001311126.1 | |||
NM_001324198.1 | 189 | Intron | NP_001311127.1 | |||
NM_031483.6 | 189 | Intron | NP_113671.3 | |||
XM_005260570.1 | 189 | Intron | XP_005260627.1 | |||
XM_011529079.1 | 189 | Intron | XP_011527381.1 | |||
XM_017028089.1 | 189 | Intron | XP_016883578.1 | |||
XM_017028090.1 | 189 | Intron | XP_016883579.1 | |||
XM_017028091.1 | 189 | Intron | XP_016883580.1 | |||
XM_017028092.1 | 189 | Intron | XP_016883581.1 |
LOC107985381 - uncharacterized LOC107985381 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017028185.1 | 189 | UTR 5 | XP_016883674.1 |