Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600698 | ||||||||||||||||||||
Literature Links: |
HMGA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HMGA2 - high mobility group AT-hook 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300918.1 | 893 | Missense Mutation | CGA,GGA | R,G 28 | NP_001287847.1 | |
NM_001300919.1 | 893 | Missense Mutation | CGA,GGA | R,G 28 | NP_001287848.1 | |
NM_003483.4 | 893 | Missense Mutation | CGA,GGA | R,G 28 | NP_003474.1 | |
NM_003484.1 | 893 | Missense Mutation | CGA,GGA | R,G 28 | NP_003475.1 | |
XM_017019989.1 | 893 | Missense Mutation | CGA,GGA | R,G 28 | XP_016875478.1 |
RPSAP52 - ribosomal protein SA pseudogene 52 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |