Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604597 MIM: 614539 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRIP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GRIP1 - glutamate receptor interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178074.1 | Intron | NP_001171545.1 | ||||
NM_021150.3 | Intron | NP_066973.2 | ||||
XM_005268754.4 | Intron | XP_005268811.1 | ||||
XM_005268757.4 | Intron | XP_005268814.1 | ||||
XM_011538093.2 | Intron | XP_011536395.1 | ||||
XM_017019098.1 | Intron | XP_016874587.1 | ||||
XM_017019099.1 | Intron | XP_016874588.1 | ||||
XM_017019100.1 | Intron | XP_016874589.1 | ||||
XM_017019101.1 | Intron | XP_016874590.1 | ||||
XM_017019102.1 | Intron | XP_016874591.1 |
HELB - DNA helicase B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |