Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114860 MIM: 164785 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPM PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CPM - carboxypeptidase M | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005502.2 | 6991 | Intron | NP_001005502.1 | |||
NM_001874.4 | 6991 | Intron | NP_001865.1 | |||
NM_198320.3 | 6991 | Intron | NP_938079.1 |
MDM2 - MDM2 proto-oncogene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145337.2 | 6991 | UTR 3 | NP_001138809.1 | |||
NM_001145339.2 | 6991 | UTR 3 | NP_001138811.1 | |||
NM_001145340.2 | 6991 | UTR 3 | NP_001138812.1 | |||
NM_001278462.1 | 6991 | UTR 3 | NP_001265391.1 | |||
NM_002392.5 | 6991 | UTR 3 | NP_002383.2 | |||
XM_005268872.4 | 6991 | UTR 3 | XP_005268929.1 | |||
XM_006719399.3 | 6991 | UTR 3 | XP_006719462.1 | |||
XM_006719400.3 | 6991 | UTR 3 | XP_006719463.1 |
Set Membership: |
HapMap |