Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610360 | ||||||||||||||||||||
Literature Links: |
LINC00969 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LINC00969 - long intergenic non-protein coding RNA 969 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MUC20 - mucin 20, cell surface associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282506.1 | 23 | UTR 5 | NP_001269435.1 | |||
NM_001291833.1 | 23 | UTR 5 | NP_001278762.1 | |||
NM_020790.1 | 23 | UTR 5 | NP_065841.1 | |||
NM_152673.3 | 23 | UTR 5 | NP_689886.3 |