Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAACAGTCATCGGATCTACAGGAC[A/C]AGTTGAATCATCTGTTGAAATAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611806 MIM: 616600 | ||||||||||||||||||||
Literature Links: |
AS3MT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AS3MT - arsenite methyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BORCS7 - BLOC-1 related complex subunit 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136200.1 | 375 | Missense Mutation | AAG,CAG | K,Q 100 | NP_001129672.1 | |
NM_144591.3 | 375 | Missense Mutation | AAG,CAG | K,Q 100 | NP_653192.2 |
BORCS7-ASMT - BORCS7-ASMT readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |