Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601568 | ||||||||||||||||||||
Literature Links: |
ADD3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADD3 - adducin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001121.3 | Intron | NP_001112.2 | ||||
NM_001320591.1 | Intron | NP_001307520.1 | ||||
NM_001320592.1 | Intron | NP_001307521.1 | ||||
NM_001320593.1 | Intron | NP_001307522.1 | ||||
NM_001320594.1 | Intron | NP_001307523.1 | ||||
NM_016824.4 | Intron | NP_058432.1 | ||||
NM_019903.4 | Intron | NP_063968.1 |
ADD3-AS1 - ADD3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |