Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
PLEKHS1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
PLEKHS1 - pleckstrin homology domain containing S1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193434.1 | Intron | NP_001180363.1 | ||||
NM_001193435.1 | Intron | NP_001180364.1 | ||||
NM_024889.4 | Intron | NP_079165.3 | ||||
NM_182601.1 | Intron | NP_872407.1 | ||||
XM_005270162.2 | Intron | XP_005270219.1 | ||||
XM_005270163.2 | Intron | XP_005270220.1 | ||||
XM_011540171.1 | Intron | XP_011538473.1 | ||||
XM_011540172.1 | Intron | XP_011538474.1 | ||||
XM_011540173.1 | Intron | XP_011538475.1 | ||||
XM_011540174.2 | Intron | XP_011538476.1 | ||||
XM_011540175.1 | Intron | XP_011538477.1 | ||||
XM_011540176.1 | Intron | XP_011538478.1 | ||||
XM_017016666.1 | Intron | XP_016872155.1 |