Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605924 | ||||||||||||||||||||
Literature Links: |
WDR4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WDR4 - WD repeat domain 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001260474.1 | Intron | NP_001247403.1 | ||||
NM_001260475.1 | Intron | NP_001247404.1 | ||||
NM_001260476.1 | Intron | NP_001247405.1 | ||||
NM_001260477.1 | Intron | NP_001247406.1 | ||||
NM_018669.5 | Intron | NP_061139.2 | ||||
NM_033661.4 | Intron | NP_387510.1 | ||||
XM_011529433.1 | Intron | XP_011527735.1 | ||||
XM_017028262.1 | Intron | XP_016883751.1 | ||||
XM_017028263.1 | Intron | XP_016883752.1 | ||||
XM_017028264.1 | Intron | XP_016883753.1 |