Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTGTCTGCGGAATCAGGGAAGAG[G/T]GAAAAGGGTCAGCCACAGAATTCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602184 | ||||||||||||||||||||
Literature Links: |
NDUFV3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NDUFV3 - NADH:ubiquinone oxidoreductase subunit V3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001503.1 | 189 | Missense Mutation | AGG,AGT | R,S 41 | NP_001001503.1 | |
NM_021075.3 | 189 | Missense Mutation | AGG,AGT | R,S 41 | NP_066553.3 | |
XM_011529586.2 | 189 | Missense Mutation | AGG,AGT | R,S 41 | XP_011527888.1 | |
XM_017028359.1 | 189 | Missense Mutation | AGG,AGT | R,S 41 | XP_016883848.1 |