Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTTGACTGCAAAGGCGATTGCAA[C/T]GAGGACCAGGAACACGGCCACCGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
21 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607178 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC00853 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC00853 - long intergenic non-protein coding RNA 853 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDZK1IP1 - PDZK1 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005764.3 | 297 | Missense Mutation | ATT,GTT | I,V 44 | NP_005755.1 |