Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123691 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRYZ PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CRYZ - crystallin zeta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130042.1 | Intron | NP_001123514.1 | ||||
NM_001130043.1 | Intron | NP_001123515.1 | ||||
NM_001134759.1 | Intron | NP_001128231.1 | ||||
NM_001889.3 | Intron | NP_001880.2 | ||||
XM_005270491.4 | Intron | XP_005270548.1 | ||||
XM_011540747.1 | Intron | XP_011539049.1 | ||||
XM_017000367.1 | Intron | XP_016855856.1 | ||||
XM_017000368.1 | Intron | XP_016855857.1 |
Set Membership: |
HapMap |