Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCACTTTCTTCATAGCGGCGACGC[C/T]GGAGGAGCCAGACATGGTGCACGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600874 | ||||||||||||||||||||
Literature Links: |
GNG5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GNG5 - G protein subunit gamma 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005274.2 | 370 | Missense Mutation | AGC,GGC | S,G 6 | NP_005265.1 |
RPF1 - ribosome production factor 1 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPATA1 - spermatogenesis associated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001310156.1 | 370 | Intron | NP_001297085.1 | |||
XM_011542514.2 | 370 | Intron | XP_011540816.1 | |||
XM_011542515.2 | 370 | Intron | XP_011540817.1 | |||
XM_011542516.1 | 370 | Intron | XP_011540818.1 | |||
XM_011542517.1 | 370 | Intron | XP_011540819.1 | |||
XM_011542519.2 | 370 | Intron | XP_011540821.1 | |||
XM_011542520.2 | 370 | Intron | XP_011540822.1 | |||
XM_011542521.2 | 370 | Intron | XP_011540823.1 | |||
XM_011542522.2 | 370 | Intron | XP_011540824.1 |