Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCTGGTCAAGGCCCACCGCAGCT[C/T]GGTCTCTGACTACGTCAACTATGAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601974 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928370 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
LOC101928370 - uncharacterized LOC101928370 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
S1PR1 - sphingosine-1-phosphate receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320730.1 | 450 | Missense Mutation | TCG,TTG | S,L 15 | NP_001307659.1 | |
NM_001400.4 | 450 | Missense Mutation | TCG,TTG | S,L 15 | NP_001391.2 |
Set Membership: |
HapMap |