Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600953 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL18 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IL18 - interleukin 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243211.1 | 793 | Silent Mutation | AAC,AAT | N,N 187 | NP_001230140.1 | |
NM_001562.3 | 793 | Silent Mutation | AAC,AAT | N,N 191 | NP_001553.1 | |
XM_011542805.1 | 793 | Silent Mutation | AAC,AAT | N,N 187 | XP_011541107.1 | |
XM_011542806.2 | 793 | Silent Mutation | AAC,AAT | N,N 191 | XP_011541108.1 |
Set Membership: |
HapMap |