Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATTCCGCCAAGCCAGATTCGGGC[A/G]CTCCCATCTTGCGCAAGTTGGTCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607854 MIM: 134770 | ||||||||||||||||||||
Literature Links: |
BEST1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BEST1 - bestrophin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001139443.1 | 717 | Intron | NP_001132915.1 | |||
NM_001300786.1 | 717 | Intron | NP_001287715.1 | |||
NM_001300787.1 | 717 | Intron | NP_001287716.1 | |||
NM_004183.3 | 717 | Intron | NP_004174.1 | |||
XM_005274210.3 | 717 | Intron | XP_005274267.1 | |||
XM_005274215.3 | 717 | Intron | XP_005274272.1 | |||
XM_005274216.3 | 717 | Intron | XP_005274273.1 | |||
XM_005274219.3 | 717 | Intron | XP_005274276.1 | |||
XM_005274221.3 | 717 | Intron | XP_005274278.1 | |||
XM_011545229.2 | 717 | Intron | XP_011543531.1 | |||
XM_011545230.2 | 717 | Intron | XP_011543532.1 | |||
XM_011545233.2 | 717 | Intron | XP_011543535.1 | |||
XM_017018228.1 | 717 | Intron | XP_016873717.1 | |||
XM_017018229.1 | 717 | Intron | XP_016873718.1 | |||
XM_017018230.1 | 717 | Intron | XP_016873719.1 |
FTH1 - ferritin heavy chain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002032.2 | 717 | Missense Mutation | GCG,GTG | A,V 161 | NP_002023.2 |
LOC399900 - uncharacterized LOC399900 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |