Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCATAGTCTGGAGTGTAGACCAG[C/G]GATCTTACAAAAGCCTCCTTGTCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604626 | ||||||||||||||||||||
Literature Links: |
ME3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ME3 - malic enzyme 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014811.1 | 1942 | Silent Mutation | TCC,TCG | S,S 577 | NP_001014811.1 | |
NM_001161586.1 | 1942 | Silent Mutation | TCC,TCG | S,S 577 | NP_001155058.1 | |
NM_006680.2 | 1942 | Silent Mutation | TCC,TCG | S,S 577 | NP_006671.2 | |
XM_005273717.1 | 1942 | Silent Mutation | TCC,TCG | S,S 577 | XP_005273774.1 | |
XM_017017136.1 | 1942 | Silent Mutation | TCC,TCG | S,S 697 | XP_016872625.1 | |
XM_017017137.1 | 1942 | Silent Mutation | TCC,TCG | S,S 440 | XP_016872626.1 | |
XM_017017138.1 | 1942 | Silent Mutation | TCC,TCG | S,S 376 | XP_016872627.1 |