Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTGACAAAGCGCCTGGCCTGTTA[C/T]GGCATTTCTTAAAAAAATAAAAATA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603845 MIM: 601926 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDUFC2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NDUFC2 - NADH:ubiquinone oxidoreductase subunit C2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204054.1 | Intron | NP_001190983.1 | ||||
NM_001204055.1 | Intron | NP_001190984.1 | ||||
NM_004549.5 | Intron | NP_004540.1 |
NDUFC2-KCTD14 - NDUFC2-KCTD14 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001203260.1 | Intron | NP_001190189.1 | ||||
NM_001203261.1 | Intron | NP_001190190.1 | ||||
NM_001203262.1 | Intron | NP_001190191.1 |
THRSP - thyroid hormone responsive | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |