Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGGCCCATGCTAGCTCACAGGC[A/G]GTGTGGGGCCTGGTGCCCTGCAACC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 152390 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALOX5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ALOX5 - arachidonate 5-lipoxygenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000698.4 | Intron | NP_000689.1 | ||||
NM_001256153.2 | Intron | NP_001243082.1 | ||||
NM_001256154.2 | Intron | NP_001243083.1 | ||||
NM_001320861.1 | Intron | NP_001307790.1 | ||||
NM_001320862.1 | Intron | NP_001307791.1 | ||||
XM_017016012.1 | Intron | XP_016871501.1 |
Set Membership: |
HapMap |