Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCTTAGAAGCAGTATTCTTTTGT[C/G]CTTAAGTGCGTTTTTGTGGCTGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616778 | ||||||||||||||||||||||||||||||||
Literature Links: |
DUSP22 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
DUSP22 - dual specificity phosphatase 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286555.1 | Intron | NP_001273484.1 | ||||
NM_020185.4 | Intron | NP_064570.1 | ||||
XM_011514757.2 | Intron | XP_011513059.1 | ||||
XM_017011061.1 | Intron | XP_016866550.1 | ||||
XM_017011062.1 | Intron | XP_016866551.1 |
Set Membership: |
HapMap |