Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGAGCGTCGGGGCCCATAAGGGA[A/G]CAGCTGATCCAAGGCCCGGTCCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606251 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GALNT9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GALNT9 - polypeptide N-acetylgalactosaminyltransferase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122636.1 | Intron | NP_001116108.1 | ||||
NM_021808.3 | Intron | NP_068580.2 | ||||
XM_017019371.1 | Intron | XP_016874860.1 | ||||
XM_017019372.1 | Intron | XP_016874861.1 | ||||
XM_017019373.1 | Intron | XP_016874862.1 | ||||
XM_017019374.1 | Intron | XP_016874863.1 | ||||
XM_017019375.1 | Intron | XP_016874864.1 | ||||
XM_017019376.1 | Intron | XP_016874865.1 |
Set Membership: |
HapMap |