Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180072 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDE6B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PDE6B - phosphodiesterase 6B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000283.3 | Intron | NP_000274.2 | ||||
NM_001145291.1 | Intron | NP_001138763.1 | ||||
NM_001145292.1 | Intron | NP_001138764.1 | ||||
XM_011513473.2 | Intron | XP_011511775.1 | ||||
XM_011513474.2 | Intron | XP_011511776.1 | ||||
XM_011513475.2 | Intron | XP_011511777.1 | ||||
XM_011513476.2 | Intron | XP_011511778.1 | ||||
XM_011513477.2 | Intron | XP_011511779.1 | ||||
XM_011513478.2 | Intron | XP_011511780.1 | ||||
XM_017008283.1 | Intron | XP_016863772.1 | ||||
XM_017008284.1 | Intron | XP_016863773.1 | ||||
XM_017008285.1 | Intron | XP_016863774.1 | ||||
XM_017008286.1 | Intron | XP_016863775.1 | ||||
XM_017008287.1 | Intron | XP_016863776.1 | ||||
XM_017008288.1 | Intron | XP_016863777.1 | ||||
XM_017008289.1 | Intron | XP_016863778.1 |
Set Membership: |
HapMap |