Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM53A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FAM53A - family with sequence similarity 53 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013622.3 | Intron | NP_001013644.1 | ||||
NM_001174070.2 | Intron | NP_001167541.1 | ||||
NM_001297435.1 | Intron | NP_001284364.1 | ||||
XM_005247948.3 | Intron | XP_005248005.1 | ||||
XM_011513402.2 | Intron | XP_011511704.1 | ||||
XM_011513403.1 | Intron | XP_011511705.1 | ||||
XM_011513404.2 | Intron | XP_011511706.1 | ||||
XM_011513405.1 | Intron | XP_011511707.1 | ||||
XM_017007802.1 | Intron | XP_016863291.1 |