Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTTTGTTGCCTCTTCTCTCTGGG[C/T]GCAGTGAGTTTCTCCCAGCCTGGGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613332 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MARCH2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
MARCH2 - membrane associated ring-CH-type finger 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005415.1 | Intron | NP_001005415.1 | ||||
NM_001005416.1 | Intron | NP_001005416.1 | ||||
NM_016496.4 | Intron | NP_057580.3 | ||||
XM_006722763.3 | Intron | XP_006722826.1 | ||||
XM_011528046.2 | Intron | XP_011526348.1 | ||||
XM_011528047.2 | Intron | XP_011526349.1 | ||||
XM_017026853.1 | Intron | XP_016882342.1 | ||||
XM_017026854.1 | Intron | XP_016882343.1 |
Set Membership: |
HapMap |