Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTATACCAGAGACGCCCATTTCTG[C/T]TCCGGGCAGATTACCTCGTGTTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
47 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100288175 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC100288175 - uncharacterized LOC100288175 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105378948 - uncharacterized LOC105378948 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF223 - ring finger protein 223 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001205252.1 | 306 | Intron | NP_001192181.1 | |||
XM_017001286.1 | 306 | Intron | XP_016856775.1 | |||
XM_017001287.1 | 306 | UTR 5 | XP_016856776.1 |