Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300383 MIM: 311040 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFP PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CFP - complement factor properdin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ELK1 - ELK1, ETS transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114123.2 | 698 | Silent Mutation | CCC,CCT | P,P 418 | NP_001107595.1 | |
NM_001257168.1 | 698 | UTR 3 | NP_001244097.1 | |||
NM_005229.4 | 698 | Silent Mutation | CCC,CCT | P,P 418 | NP_005220.2 | |
XM_017029339.1 | 698 | Silent Mutation | CCC,CCT | P,P 418 | XP_016884828.1 |
Set Membership: |
HapMap |