Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCGCTCCCGGCGGGGACAGGCTG[G/T]CCGCCTCCAGCTCGGCTGCCAGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 142992 | ||||||||||||||||||||
Literature Links: |
HMX1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HMX1 - H6 family homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306142.1 | 1009 | Intron | NP_001293071.1 | |||
NM_018942.2 | 1009 | Missense Mutation | GAC,GCC | D,A 269 | NP_061815.2 |