Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCAATCTGGTAGATGAACCACAC[A/G]GCCAGCTTTTGGGTGCAGGCACAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD18B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANKRD18B - ankyrin repeat domain 18B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244752.1 | Intron | NP_001231681.1 | ||||
XM_011517881.2 | Intron | XP_011516183.1 | ||||
XM_011517883.2 | Intron | XP_011516185.1 | ||||
XM_011517885.2 | Intron | XP_011516187.1 | ||||
XM_017014728.1 | Intron | XP_016870217.1 |