Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605477 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARHGEF7 - Rho guanine nucleotide exchange factor 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEX29 - testis expressed 29 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303133.1 | Intron | NP_001290062.1 | ||||
NM_152324.2 | Intron | NP_689537.1 | ||||
XM_017020387.1 | Intron | XP_016875876.1 | ||||
XM_017020388.1 | Intron | XP_016875877.1 |