Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCAGATCACTCACTGCCCAGCA[A/G]TGGGGGCTTGGGGACCCCCACTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
48 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176873 MIM: 611141 MIM: 603321 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK11B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDK11B - cyclin dependent kinase 11B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIB2 - mindbomb E3 ubiquitin protein ligase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MMP23B - matrix metallopeptidase 23B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006983.1 | 147 | Intron | NP_008914.1 | |||
XM_017002615.1 | 147 | UTR 5 | XP_016858104.1 | |||
XM_017002616.1 | 147 | UTR 5 | XP_016858105.1 | |||
XM_017002617.1 | 147 | UTR 5 | XP_016858106.1 |