Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116951 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK11A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDK11A - cyclin dependent kinase 11A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001313896.1 | Intron | NP_001300825.1 | ||||
NM_001313982.1 | Intron | NP_001300911.1 | ||||
NM_024011.3 | Intron | NP_076916.2 | ||||
NM_033529.3 | Intron | NP_277071.2 |
SLC35E2 - solute carrier family 35 member E2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |