Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGCCTCCTTTGTGATGCAGGCA[G/T]GGGCTCCCTGGAGGCGGTGACTCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
MMEL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MMEL1 - membrane metallo-endopeptidase-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033467.3 | Intron | NP_258428.2 | ||||
XM_011542122.2 | Intron | XP_011540424.1 | ||||
XM_017002310.1 | Intron | XP_016857799.1 | ||||
XM_017002311.1 | Intron | XP_016857800.1 | ||||
XM_017002312.1 | Intron | XP_016857801.1 | ||||
XM_017002313.1 | Intron | XP_016857802.1 | ||||
XM_017002314.1 | Intron | XP_016857803.1 | ||||
XM_017002315.1 | Intron | XP_016857804.1 |
TTC34 - tetratricopeptide repeat domain 34 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |