Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGCCGGGGGAAGGTCGGCGGCCG[C/G]GTCGGCAGGCGGGAGGGCGGAGTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608060 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HES4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HES4 - hes family bHLH transcription factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142467.1 | 313 | Intron | NP_001135939.1 | |||
NM_021170.3 | 313 | Intron | NP_066993.1 | |||
XM_005244771.4 | 313 | UTR 5 | XP_005244828.1 |