Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGTCCCCGTGAGGGCCGCCAGGT[C/T]GGCTGTAGCCTTTACCACAGGCCAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 103320 MIM: 147571 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGRN PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AGRN - agrin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305275.1 | Intron | NP_001292204.1 | ||||
NM_198576.3 | Intron | NP_940978.2 | ||||
XM_005244749.3 | Intron | XP_005244806.1 | ||||
XM_011541429.2 | Intron | XP_011539731.1 |
ISG15 - ISG15 ubiquitin-like modifier | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |