Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
28 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 103320 MIM: 147571 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGRN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AGRN - agrin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305275.1 | Intron | NP_001292204.1 | ||||
NM_198576.3 | Intron | NP_940978.2 | ||||
XM_005244749.3 | Intron | XP_005244806.1 | ||||
XM_011541429.2 | Intron | XP_011539731.1 |
ISG15 - ISG15 ubiquitin-like modifier | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |