Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAACATGCTAATAGAAGAGCAGCC[T/C]CAGACTCACTGCCTGTCCAGGTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
17 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614264 MIM: 616041 MIM: 191523 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP30 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGAP30 - Rho GTPase activating protein 30 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSTD1 - thiosulfate sulfurtransferase like domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USF1 - upstream transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276373.1 | Intron | NP_001263302.1 | ||||
NM_007122.4 | Intron | NP_009053.1 | ||||
NM_207005.2 | Intron | NP_996888.1 |