Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCGCCGTCTTCCTGTCGGCGGCC[A/G]GCACTGTAGGTGAGCGCGAGAGGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611772 MIM: 603276 | ||||||||||||||||||||
Literature Links: |
NUF2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NUF2 - NUF2, NDC80 kinetochore complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031423.3 | 125 | UTR 5 | NP_113611.2 | |||
NM_145697.2 | 125 | UTR 5 | NP_663735.2 | |||
XM_011510036.1 | 125 | UTR 5 | XP_011508338.1 |
RGS5 - regulator of G-protein signaling 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195303.2 | 125 | Intron | NP_001182232.1 | |||
NM_001254748.1 | 125 | Intron | NP_001241677.1 | |||
NM_001254749.1 | 125 | Intron | NP_001241678.1 | |||
NM_003617.3 | 125 | Intron | NP_003608.1 |