Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACCTTTTAAGGGTTATGATTCAG[G/T]AGATCTTTCATAGGGCCTAGAAATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611541 | ||||||||||||||||||||
Literature Links: |
SNX27 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SNX27 - sorting nexin family member 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030918.5 | Intron | NP_112180.4 | ||||
XM_005245509.2 | Intron | XP_005245566.1 | ||||
XM_005245510.3 | Intron | XP_005245567.1 | ||||
XM_005245511.4 | Intron | XP_005245568.1 | ||||
XM_011510024.2 | Intron | XP_011508326.1 | ||||
XM_011510025.2 | Intron | XP_011508327.1 | ||||
XM_011510026.2 | Intron | XP_011508328.1 | ||||
XM_017002417.1 | Intron | XP_016857906.1 |