Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGTGTCTACAGGTGTTTTAGTTG[C/T]GAATCTTTATTTCTTCTGCTTACTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 136132 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FMO3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FMO3 - flavin containing monooxygenase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002294.2 | Intron | NP_001002294.1 | ||||
NM_001319173.1 | Intron | NP_001306102.1 | ||||
NM_001319174.1 | Intron | NP_001306103.1 | ||||
NM_006894.5 | Intron | NP_008825.4 | ||||
XM_011509345.2 | Intron | XP_011507647.1 |
MIR1295A - microRNA 1295a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR1295B - microRNA 1295b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |