Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACATTTGAGTAAATGAGGAGCTG[C/G]TGCTAGAGGACAGAAATCCCAGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 136130 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FMO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FMO1 - flavin containing monooxygenase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282692.1 | Intron | NP_001269621.1 | ||||
NM_001282693.1 | Intron | NP_001269622.1 | ||||
NM_001282694.1 | Intron | NP_001269623.1 | ||||
NM_002021.2 | Intron | NP_002012.1 | ||||
XM_005245037.3 | Intron | XP_005245094.1 | ||||
XM_005245038.3 | Intron | XP_005245095.1 | ||||
XM_006711241.3 | Intron | XP_006711304.1 | ||||
XM_006711242.3 | Intron | XP_006711305.1 |