Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTAAAGGGAGGAAGAATACTTTC[A/G]TCTTGCCAAGAGCTCCAAGAGGAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606820 MIM: 600063 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLRX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLRX2 - glutaredoxin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243399.1 | Intron | NP_001230328.1 | ||||
NM_001319291.1 | Intron | NP_001306220.1 | ||||
NM_016066.4 | Intron | NP_057150.2 | ||||
NM_197962.2 | Intron | NP_932066.1 | ||||
XM_017001397.1 | Intron | XP_016856886.1 |
TROVE2 - TROVE domain family member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |