Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAGCAGGTCCAGTTCCACCACTT[C/T]ATAGTTAACATTCATGTCATGGAAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606820 MIM: 600063 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLRX2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GLRX2 - glutaredoxin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243399.1 | 604 | Missense Mutation | AAA,GAA | K,E 55 | NP_001230328.1 | |
NM_001319291.1 | 604 | Missense Mutation | AAA,GAA | K,E 55 | NP_001306220.1 | |
NM_016066.4 | 604 | Missense Mutation | AAA,GAA | K,E 96 | NP_057150.2 | |
NM_197962.2 | 604 | Missense Mutation | AAA,GAA | K,E 95 | NP_932066.1 | |
XM_017001397.1 | 604 | Missense Mutation | AAA,GAA | K,E 55 | XP_016856886.1 |
TROVE2 - TROVE domain family member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |