Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGATGGCCTCAGGAGCAAGCAGGG[C/G]TCCCTTGGCTGCCTCTGCTGCTGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 147880 | ||||||||||||||||||||
Literature Links: |
IL6R PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IL6R - interleukin 6 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000565.3 | Intron | NP_000556.1 | ||||
NM_001206866.1 | Intron | NP_001193795.1 | ||||
NM_181359.2 | Intron | NP_852004.1 | ||||
XM_005245139.1 | Intron | XP_005245196.1 | ||||
XM_005245140.2 | Intron | XP_005245197.1 | ||||
XM_006711298.1 | Intron | XP_006711361.1 | ||||
XM_006711299.3 | Intron | XP_006711362.1 | ||||
XM_017001199.1 | Intron | XP_016856688.1 | ||||
XM_017001200.1 | Intron | XP_016856689.1 | ||||
XM_017001201.1 | Intron | XP_016856690.1 |
LOC101928101 - uncharacterized LOC101928101 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |