Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTAAGTCATGATTATCTGAGCAGA[G/T]ATGATTTTATACCCTCTAGGATTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 146920 MIM: 118507 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAR - adenosine deaminase, RNA specific | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025107.2 | 4672 | UTR 3 | NP_001020278.1 | |||
NM_001111.4 | 4672 | UTR 3 | NP_001102.2 | |||
NM_001193495.1 | 4672 | UTR 3 | NP_001180424.1 | |||
NM_015840.3 | 4672 | UTR 3 | NP_056655.2 | |||
NM_015841.3 | 4672 | UTR 3 | NP_056656.2 | |||
XM_006711109.2 | 4672 | UTR 3 | XP_006711172.1 | |||
XM_006711111.3 | 4672 | UTR 3 | XP_006711174.1 | |||
XM_006711112.2 | 4672 | UTR 3 | XP_006711175.1 | |||
XM_006711113.2 | 4672 | UTR 3 | XP_006711176.1 | |||
XM_011509060.1 | 4672 | UTR 3 | XP_011507362.1 | |||
XM_011509061.2 | 4672 | UTR 3 | XP_011507363.2 | |||
XM_011509062.1 | 4672 | UTR 3 | XP_011507364.1 | |||
XM_017000037.1 | 4672 | UTR 3 | XP_016855526.1 |
CHRNB2 - cholinergic receptor nicotinic beta 2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000748.2 | 4672 | Intron | NP_000739.1 |