Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGAAAGCCTATAGCTATTCTTAC[A/G]AAAAGCACCATTTCAGCCAGGAACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PM20D1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PM20D1 - peptidase M20 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152491.4 | Intron | NP_689704.4 | ||||
XM_017000396.1 | Intron | XP_016855885.1 | ||||
XM_017000397.1 | Intron | XP_016855886.1 | ||||
XM_017000398.1 | Intron | XP_016855887.1 |