Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGTTCGGTGGAAATCAGCCGTAG[C/T]CATGAGTTTCTGCCGGGGCTAGCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 179755 | ||||||||||||||||||||
Literature Links: |
PRCC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PRCC - papillary renal cell carcinoma (translocation-associated) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005973.4 | 134 | UTR 5 | NP_005964.3 | |||
XM_005245313.2 | 134 | UTR 5 | XP_005245370.1 | |||
XM_005245314.1 | 134 | UTR 5 | XP_005245371.1 | |||
XM_011509726.1 | 134 | UTR 5 | XP_011508028.1 |