Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
JMJD4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
JMJD4 - jumonji domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161465.1 | 2438 | UTR 3 | NP_001154937.1 | |||
NM_023007.2 | 2438 | UTR 3 | NP_075383.2 |
SNAP47 - synaptosome associated protein 47 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323930.1 | 2438 | Intron | NP_001310859.1 | |||
NM_001323931.1 | 2438 | Intron | NP_001310860.1 | |||
NM_001323932.1 | 2438 | Intron | NP_001310861.1 | |||
NM_001323933.1 | 2438 | Intron | NP_001310862.1 | |||
NM_001323934.1 | 2438 | Intron | NP_001310863.1 | |||
NM_001323935.1 | 2438 | Intron | NP_001310864.1 | |||
NM_053052.3 | 2438 | Intron | NP_444280.2 |