Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTAATCACTAGACCGGGTATCCT[C/T]TATAGCACAGTTTTATCCTGCTGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605454 | ||||||||||||||||||||
Literature Links: |
ABCB10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABCB10 - ATP binding cassette subfamily B member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012089.2 | Intron | NP_036221.2 | ||||
XM_011544135.2 | Intron | XP_011542437.1 | ||||
XM_011544136.2 | Intron | XP_011542438.1 |
LOC101927478 - uncharacterized LOC101927478 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |