Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCTTGCTGAATGAGTGATGTGAT[A/G]GGTAGCACCATTTTGGCCCATAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179836 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RPA2 - replication protein A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286076.1 | Intron | NP_001273005.1 | ||||
NM_001297558.1 | Intron | NP_001284487.1 | ||||
NM_002946.4 | Intron | NP_002937.1 |
THEMIS2 - thymocyte selection associated family member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |