Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATACATGAGAATGTATTCTTTTA[A/G]GAGATTTTTCACATAGTGGTCAACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 604409 | ||||||||||||||||||||
Literature Links: |
GMEB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GMEB1 - glucocorticoid modulatory element binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319674.1 | Intron | NP_001306603.1 | ||||
NM_006582.3 | Intron | NP_006573.2 | ||||
NM_024482.2 | Intron | NP_077808.1 | ||||
XM_011540518.2 | Intron | XP_011538820.1 | ||||
XM_011540519.2 | Intron | XP_011538821.1 | ||||
XM_011540521.2 | Intron | XP_011538823.1 | ||||
XM_011540522.2 | Intron | XP_011538824.1 | ||||
XM_017000087.1 | Intron | XP_016855576.1 |